The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006451.4 | NP_006442.2 | 11 - 11 | 2559 | 125 |
NM_182789.3 | NP_877590.1 | 11 - 11 | 2322 | 125 | |
NM_183323.2 | NP_899152.1 | 11 - 11 | 2127 | 125 | |
XM_005248230.3 | XP_005248287.1 | 11 - 11 | 2281 | 125 | |
XM_017008956.1 | XP_016864445.1 | 11 - 11 | 2462 | 125 | |
GenBank mRNA | AL136920.1 | - | 11 - 11 | 2508 | 125 |
DQ786248.1 | - | 1 - 1 | 815 | 125 |