The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001043351.1 | NP_001036816.1 | 8 - 8 | 1240 | 118 |
NM_001043352.1 | NP_001036817.1 | 8 - 8 | 2567 | 118 | |
NM_001043353.1 | NP_001036818.1 | 8 - 8 | 2567 | 118 | |
NM_001278188.1 | NP_001265117.1 | 7 - 7 | 1106 | 118 | |
NM_001278189.1 | NP_001265118.1 | 9 - 9 | 1319 | 118 | |
NM_001278190.1 | NP_001265119.1 | 7 - 7 | 1177 | 118 | |
NM_001278191.1 | NP_001265120.1 | 8 - 8 | 1086 | 118 | |
NM_153649.3 | NP_705935.1 | 8 - 8 | 1240 | 118 | |
NR_103461.1 | 8 - 8 | 1236 | 118 | ||
GenBank mRNA | AB062125.1 | - | 4 - 4 | 653 | 118 |
AK026559.1 | - | 8 - 8 | 1180 | 118 | |
AK056889.1 | - | 4 - 4 | 1229 | 118 | |
AK056921.1 | - | 8 - 8 | 1097 | 118 | |
AK092712.1 | - | 7 - 7 | 985 | 118 | |
AK123113.1 | - | 1 - 1 | 1362 | 118 | |
AK125367.1 | - | 5 - 5 | 3124 | 118 | |
AK298678.1 | - | 7 - 7 | 1084 | 118 | |
BC000771.2 | - | 8 - 8 | 1152 | 118 | |
BC015403.1 | - | 8 - 8 | 1155 | 118 | |
BC072428.1 | - | 8 - 8 | 1167 | 118 | |
BX641075.1 | - | 1 - 1 | 3870 | 118 | |
X04588.1 | - | 8 - 8 | 1160 | 118 |