Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR7D2
Entrez Gene ID
162998
Gene Name
olfactory receptor family 7 subfamily D member 2
Gene Aliases
HTPCRH03, OR19-10, OR19-4
Chromosome Location
Chr.19: 9185594 - 9188817 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175883.2 NP_787079.1 1 - 1 630 68
GenBank mRNA AK095468.1 - 1 - 1 630 68
X64975.1 - 1 - 1 68 68

View Full Product Details