The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000514.3 | NP_000505.1 | 3 - 3 | 742 | 91 |
NM_001190468.1 | NP_001177397.1 | 3 - 3 | 762 | 91 | |
NM_001190469.1 | NP_001177398.1 | 3 - 3 | 684 | 91 | |
NM_001278098.1 | NP_001265027.1 | 3 - 3 | 570 | 91 | |
NM_199231.2 | NP_954701.1 | 3 - 3 | 587 | 91 | |
XM_011514030.2 | XP_011512332.1 | 2 - 2 | 455 | 91 | |
XM_017009337.1 | XP_016864826.1 | 3 - 3 | 1028 | 91 | |
GenBank mRNA | AY052832.1 | - | 2 - 2 | 464 | 91 |
BC069119.1 | - | 2 - 2 | 542 | 91 | |
BC069369.1 | - | 2 - 2 | 542 | 91 | |
BC128108.1 | - | 2 - 2 | 542 | 91 | |
BC128109.1 | - | 2 - 2 | 542 | 91 | |
CR541923.1 | - | 2 - 2 | 542 | 91 | |
DQ235474.1 | - | 3 - 3 | 544 | 91 | |
KU178093.1 | - | 2 - 2 | 542 | 91 | |
KU178094.1 | - | 2 - 2 | 464 | 91 |