Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COMMD6
Entrez Gene ID
170622
Gene Name
COMM domain containing 6
Gene Aliases
Acrg
Chromosome Location
Chr.13: 75525214 - 75549439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287392.1 NP_001274321.1 4 - 4 1022 145
NM_001287393.1 NP_001274322.1 4 - 4 1007 145
NM_001287394.1 NP_001274323.1 3 - 3 838 145
NM_203495.3 NP_987091.1 4 - 4 842 145
NM_203497.3 NP_987093.1 5 - 5 881 145
GenBank mRNA AF086081.1 - 1 - 1 465 145
BC110837.1 - 4 - 4 779 145
BX106312.1 - 1 - 1 111 145
DB574884.1 - 1 - 1 378 145

View Full Product Details