The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020957.3 | NP_066008.2 | 1 - 1 | 1886 | 158 |
GenBank mRNA | AB046841.1 | - | 1 - 1 | 2173 | 158 |
AF217757.1 | - | 1 - 1 | 1016 | 158 | |
AK056460.1 | - | 1 - 1 | 1191 | 158 | |
AY013878.1 | - | 1 - 1 | 1016 | 158 | |
BC036062.1 | - | 1 - 1 | 1165 | 158 |