The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201329.1 | NP_001188258.1 | 2 - 2 | 3898 | 144 |
NM_024607.3 | NP_078883.2 | 2 - 2 | 3750 | 144 | |
XM_006716253.3 | XP_006716316.1 | 2 - 2 | 3703 | 144 | |
XM_017013835.1 | XP_016869324.1 | 2 - 2 | 3850 | 144 | |
GenBank mRNA | AK024067.1 | - | 2 - 2 | 3736 | 144 |