The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098515.1 | NP_001091985.1 | 3 - 3 | 2196 | 165 |
NM_145015.4 | NP_659452.3 | 3 - 3 | 2185 | 165 | |
XM_017017170.1 | XP_016872659.1 | 3 - 3 | 2694 | 165 | |
GenBank mRNA | AK075450.1 | - | 3 - 3 | 2071 | 165 |
AK075492.1 | - | 4 - 4 | 1536 | 165 | |
AK098317.1 | - | 3 - 3 | 2006 | 165 | |
AK131068.1 | - | 2 - 2 | 1788 | 165 | |
AL049451.1 | - | 1 - 1 | 724 | 165 | |
BC016964.1 | - | 3 - 3 | 2003 | 165 |