The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175887.2 | NP_787083.1 | 2 - 2 | 1551 | 156 |
XM_005249653.3 | XP_005249710.1 | 2 - 2 | 2354 | 156 | |
XM_011515198.1 | XP_011513500.1 | 2 - 2 | 2349 | 156 | |
XM_011515199.1 | XP_011513501.1 | 2 - 2 | 1263 | 156 | |
XM_017011842.1 | XP_016867331.1 | 2 - 2 | 1561 | 156 | |
GenBank mRNA | BC018144.1 | - | 2 - 2 | 1447 | 156 |
BC029131.1 | - | 2 - 2 | 1551 | 156 | |
FJ857942.1 | - | 1 - 1 | 870 | 156 |