The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001715.3 | NP_001001715.2 | 3 - 3 | 1150 | 154 |
GenBank mRNA | BE463494.1 | - | 1 - 1 | 154 | |
BQ014327.1 | - | 1 - 1 | 154 |