Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR5W2
Entrez Gene ID
390148
Gene Name
olfactory receptor family 5 subfamily W member 2
Gene Aliases
OR5W2P, OR5W3P
Chromosome Location
Chr.11: 55913650 - 55914582 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001960.1 NP_001001960.1 1 - 1 869 129

View Full Product Details