Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR2T33
Entrez Gene ID
391195
Gene Name
olfactory receptor family 2 subfamily T member 33
Gene Aliases
OR1-56
Chromosome Location
Chr.1: 248272852 - 248273814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004695.1 NP_001004695.1 1 - 1 911 127
GenBank mRNA BC136994.1 - 1 - 1 933 127
BC136995.1 - 1 - 1 933 127

View Full Product Details