Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR5AN1
Entrez Gene ID
390195
Gene Name
olfactory receptor family 5 subfamily AN member 1
Gene Aliases
OR11-244
Chromosome Location
Chr.11: 59364459 - 59365394 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004729.1 NP_001004729.1 1 - 1 857 135
GenBank mRNA BC140917.1 - 1 - 1 135

View Full Product Details