Product Details

Assay Reference Genome
Location

Chr.17:7687604 on build GRCh38
Cytoband
17p13.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55135
Gene Symbol
WRAP53
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
Location
Chr.17:7686071-7703502 on build GRCh38
Assay Gene Location
Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
WRAP53 NM_001143990.1 NP_001137462.1
NM_001143991.1 NP_001137463.1
XM_017024812.1 1 33 XP_016880301.1
AK056669.1 1 925
AK092370.1
AY766322.1
DQ431241.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv574322 Chr17:7010333 - 7689462 on Build GRCh38 Loss PLSCR3 TNK1 SENP3 POLR2A CD68 ALOX12 DLG4 LOC100996842 TMEM102 CTDNEP1 NEURL4 BCL6B SPEM1 TMEM256 RNASEK-C17orf49 TNFSF12-TNFSF13 SLC35G6 C17orf49 SHBG MIR497 RNASEK ELP5 NLGN2 TNFSF12 MIR497HG WRAP53 MIR195 C17orf74 SLC16A11 ALOX12-AS1 SNORA67 ACAP1 SLC16A13 SOX15 SNORD10 KCTD11 TNFSF13 DVL2 CHRNB1 SENP3-EIF4A1 SAT2 FGF11 PHF23 ASGR1 TP53 TMEM256-PLSCR3 GPS2 CLDN7 EIF5A TMEM95 CLEC10A ASGR2 MIR324 GABARAP SLC2A4 MPDU1 ZBTB4 EIF4A1 FXR2 YBX2 SNORA48 LOC107983988 ATP1B2 ACADVL

View Full Product Details