Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
OPA3 | NM_001017989.2 | 2 | 974 | NP_001017989.2 |
AK098798.1 | 2 | 952 | BAC05415.1 | |
BC047316.1 | 2 | 895 | AAH47316.1 | |
BC064146.1 | 2 | 886 | AAH64146.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833848 | Chr19:45457372 - 45644939 on Build GRCh38 | Loss | EML2 RTN2 PPM1N GPR4 OPA3 FOSB VASP ERCC1 EML2-AS1 MIR330 |