The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007278.2 | NP_001007279.1 | 4 - 4 | 1358 | 78 |
NM_005798.4 | NP_005789.2 | 3 - 3 | 1159 | 78 | |
NM_052811.3 | NP_434698.1 | 3 - 3 | 1244 | 78 | |
NM_213590.2 | NP_998755.1 | 2 - 2 | 1694 | 78 | |
GenBank mRNA | AF220127.1 | - | 3 - 3 | 1099 | 78 |
AF220128.1 | - | 1 - 1 | 937 | 78 | |
AF241850.1 | - | 3 - 3 | 1117 | 78 | |
AJ224819.1 | - | 2 - 2 | 1044 | 78 | |
AK001806.1 | - | 1 - 1 | 385 | 78 | |
AK314496.1 | - | 3 - 3 | 1203 | 78 | |
AL360196.1 | - | 3 - 3 | 1099 | 78 | |
AY191002.1 | - | 3 - 3 | 1192 | 78 | |
AY455758.1 | - | 2 - 2 | 1669 | 78 | |
AY764035.1 | - | 4 - 4 | 1328 | 78 | |
BC003579.1 | - | 3 - 3 | 1205 | 78 | |
BC063407.1 | - | 2 - 2 | 1636 | 78 |