The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004821.2 | NP_004812.1 | 2 - 2 | 1638 | 65 |
XM_005268531.1 | XP_005268588.1 | 2 - 2 | 1580 | 65 | |
GenBank mRNA | AF061756.1 | - | 2 - 2 | 1639 | 65 |
BC021190.2 | - | 2 - 2 | 1607 | 65 |