Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR7G3
Entrez Gene ID
390883
Gene Name
olfactory receptor family 7 subfamily G member 3
Gene Aliases
OST085
Chromosome Location
Chr.19: 9126012 - 9126950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001958.1 NP_001001958.1 1 - 1 879 110

View Full Product Details