Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR13H1
Entrez Gene ID
347468
Gene Name
olfactory receptor family 13 subfamily H member 1
Gene Aliases
ORX1
Chromosome Location
Chr.X: 131544074 - 131545000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004486.1 NP_001004486.1 1 - 1 741 158
GenBank mRNA BC137050.1 - 1 - 1 839 158

View Full Product Details