Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4N2
Entrez Gene ID
390429
Gene Name
olfactory receptor family 4 subfamily N member 2
Gene Aliases
OR14-13, OR14-8
Chromosome Location
Chr.14: 19803770 - 19828372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004723.2 NP_001004723.1 2 - 2 837 170

Gene Symbol
OR4N3P
Entrez Gene ID
390539
Gene Name
olfactory receptor family 4 subfamily N member 3 pseudogene
Gene Aliases
OR15-2
Chromosome Location
Chr.15: 22125511 - 22126447 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028067.1 1 - 1 753 170
GenBank mRNA BX114189.1 - 1 - 1 299 170

View Full Product Details