The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005171.2 | NP_001005171.2 | 1 - 1 | 704 | 148 |
GenBank mRNA | BC137056.1 | - | 1 - 1 | 704 | 148 |
BC137059.1 | - | 1 - 1 | 704 | 148 |