The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002920.1 | NP_001002920.1 | 2 - 3 | 462 | 160 |
NM_001005365.2 | NP_001005365.2 | 2 - 3 | 462 | 160 | |
XM_011544516.1 | XP_011542818.1 | 2 - 3 | 534 | 160 | |
GenBank mRNA | AY462869.1 | - | 2 - 3 | 462 | 160 |
AY462870.1 | - | 2 - 3 | 462 | 160 |