Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR5K3
Entrez Gene ID
403277
Gene Name
olfactory receptor family 5 subfamily K member 3
Gene Aliases
-
Chromosome Location
Chr.3: 98390666 - 98391631 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005516.1 NP_001005516.1 1 - 1 686 150

View Full Product Details