Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP5-1
Entrez Gene ID
387264
Gene Name
keratin associated protein 5-1
Gene Aliases
KRN1L, KRTAP5.1
Chromosome Location
Chr.11: 1584342 - 1585283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005922.1 NP_001005922.1 1 - 1 898 78
GenBank mRNA AB126070.1 - 1 - 1 898 78

View Full Product Details