The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257370.1 | NP_001244299.1 | 2 - 3 | 387 | 173 |
XM_017023025.1 | XP_016878514.1 | 2 - 3 | 309 | 173 | |
GenBank mRNA | AK074080.2 | - | 2 - 3 | 289 | 173 |
EF452422.1 | - | 2 - 3 | 387 | 173 |