Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HLA-DRB5
Entrez Gene ID
3127
Gene Name
major histocompatibility complex, class II, DR beta 5
Gene Aliases
HLA-DRB
Chromosome Location
Chr.6: 32517343 - 32530316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002125.3 NP_002116.2 1 - 2 104 126
XM_011514562.2 XP_011512864.1 1 - 2 191 126
GenBank mRNA AK314834.1 - 1 - 2 146 126
BC009234.2 - 1 - 2 142 126
M20429.1 - 1 - 2 104 126
M57648.1 - 1 - 2 157 126

View Full Product Details