The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003494.1 | 4 - 5 | 422 | 151 | |
XM_006716262.1 | XP_006716325.1 | 4 - 5 | 298 | 151 | |
GenBank mRNA | AK098138.1 | - | 4 - 5 | 389 | 151 |
AK308513.1 | - | 3 - 4 | 406 | 151 | |
BC098427.1 | - | 4 - 5 | 472 | 151 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005273607.2 | XP_005273664.1 | 4 - 5 | 434 | 151 |
GenBank mRNA | AK291001.1 | - | 4 - 5 | 434 | 151 |
AK297741.1 | - | 3 - 4 | 320 | 151 | |
AK298940.1 | - | 2 - 3 | 296 | 151 | |
AK309984.1 | - | 5 - 6 | 532 | 151 |