Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC34A3
Entrez Gene ID
142680
Gene Name
solute carrier family 34 member 3
Gene Aliases
HHRH, NPTIIc
Chromosome Location
Chr.9: 137230757 - 137236554 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177316.1 NP_001170787.1 3 - 4 403 132
NM_001177317.1 NP_001170788.1 3 - 4 406 132
NM_080877.2 NP_543153.1 3 - 4 368 132
XM_011518257.2 XP_011516559.1 3 - 4 344 132
XM_011518258.1 XP_011516560.1 2 - 3 234 132
XM_011518259.1 XP_011516561.1 3 - 4 368 132
XM_011518260.1 XP_011516562.1 3 - 4 286 132
XM_011518261.2 XP_011516563.1 3 - 4 359 132
XM_017014290.1 XP_016869779.1 4 - 5 454 132
XM_017014291.1 XP_016869780.1 3 - 4 344 132
XM_017014292.1 XP_016869781.1 3 - 4 344 132
GenBank mRNA AB055000.1 - 3 - 4 252 132
AK095999.1 - 3 - 4 368 132
AL832449.1 - 3 - 4 1383 132
BC029942.1 - 3 - 4 406 132

View Full Product Details