Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C17orf58
Entrez Gene ID
284018
Gene Name
chromosome 17 open reading frame 58
Gene Aliases
-
Chromosome Location
Chr.17: 67991099 - 67993649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181655.3 NP_858041.2 2 - 3 415 107
GenBank mRNA BC036760.1 - 2 - 3 290 107
BC105124.1 - 2 - 3 234 107
BC112073.1 - 2 - 3 246 107

View Full Product Details