Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHG7
Entrez Gene ID
440107
Gene Name
pleckstrin homology and RhoGEF domain containing G7
Gene Aliases
-
Chromosome Location
Chr.12: 92736489 - 92772092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004330.2 NP_001004330.1 10 - 11 1163 128
GenBank mRNA AK128530.1 - 10 - 11 1117 128
BC137086.1 - 9 - 10 1044 128

View Full Product Details