Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRG2
Entrez Gene ID
448831
Gene Name
FSHD region gene 2
Gene Aliases
FRG2A
Chromosome Location
Chr.4: 190024367 - 190027257 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005217.1 NP_001005217.1 1 - 2 230 116
GenBank mRNA AY714545.1 - 1 - 2 230 116

Gene Symbol
FRG2B
Entrez Gene ID
441581
Gene Name
FSHD region gene 2 family member B
Gene Aliases
-
Chromosome Location
Chr.10: 133625099 - 133626795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080998.1 NP_001074467.1 1 - 2 230 116
GenBank mRNA AY744466.1 - 1 - 2 230 116

View Full Product Details