The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031308.3 | NP_112598.3 | 2 - 2 | 6695 | 180 |
XM_017013889.1 | XP_016869378.1 | 2 - 2 | 6727 | 180 | |
XM_017013890.1 | XP_016869379.1 | 2 - 2 | 6866 | 180 | |
XM_017013891.1 | XP_016869380.1 | 3 - 3 | 3844 | 180 | |
XM_017013892.1 | XP_016869381.1 | 2 - 2 | 6727 | 180 | |
GenBank mRNA | AB051895.1 | - | 1 - 1 | 6636 | 180 |