The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004854.4 | NP_004845.1 | 7 - 7 | 2393 | 63 |
XM_011512207.1 | XP_011510509.1 | 7 - 7 | 2217 | 63 | |
XM_011512208.1 | XP_011510510.1 | 7 - 7 | 2402 | 63 | |
XM_011512210.1 | XP_011510512.1 | 8 - 8 | 2457 | 63 | |
XM_011512211.1 | XP_011510513.1 | 8 - 8 | 2299 | 63 | |
XM_011512212.1 | XP_011510514.1 | 8 - 8 | 2519 | 63 | |
XM_017005380.1 | XP_016860869.1 | 9 - 9 | 2903 | 63 | |
XM_017005381.1 | XP_016860870.1 | 9 - 9 | 2717 | 63 | |
XM_017005382.1 | XP_016860871.1 | 9 - 9 | 2779 | 63 | |
XM_017005383.1 | XP_016860872.1 | 9 - 9 | 2720 | 63 | |
GenBank mRNA | AF033827.1 | - | 7 - 7 | 2381 | 63 |
AF070594.1 | - | 6 - 6 | 2069 | 63 | |
BC010441.1 | - | 7 - 7 | 2334 | 63 |