The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098672.1 | NP_001092142.1 | 18 - 19 | 3372 | 70 |
GenBank mRNA | BX641008.1 | - | 7 - 8 | 1127 | 70 |