The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003610.1 | 19 - 20 | 1896 | 129 | |
GenBank mRNA | AL833251.1 | - | 19 - 20 | 1896 | 129 |