Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP90AB2P
Entrez Gene ID
391634
Gene Name
heat shock protein 90 alpha family class B member 2, pseudogene
Gene Aliases
HSP90BB
Chromosome Location
Chr.4: 13336387 - 13338931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY956763.1 - 2 - 3 450 99

View Full Product Details