Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPIP5K1
Entrez Gene ID
9677
Gene Name
diphosphoinositol pentakisphosphate kinase 1
Gene Aliases
HISPPD2A, IP6K, IPS1, VIP1, hsVIP1
Chromosome Location
Chr.15: 43533462 - 43590272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130858.2 NP_001124330.1 26 - 27 3265 79
NM_001130859.2 NP_001124331.1 25 - 26 3248 79
NM_014659.5 NP_055474.3 25 - 26 3171 79
XM_005254803.1 XP_005254860.1 26 - 27 3253 79
XM_005254804.1 XP_005254861.1 26 - 27 3253 79
XM_011522249.1 XP_011520551.1 27 - 28 3403 79
XM_011522250.1 XP_011520552.1 27 - 28 3506 79
XM_011522252.1 XP_011520554.1 26 - 27 3377 79
XM_011522254.1 XP_011520556.1 26 - 27 3273 79
XM_011522258.1 XP_011520560.1 21 - 22 2663 79
XM_011522259.1 XP_011520561.1 26 - 27 3229 79
XM_017022735.1 XP_016878224.1 26 - 27 3720 79
XM_017022736.1 XP_016878225.1 26 - 27 3688 79
XM_017022737.1 XP_016878226.1 26 - 27 3369 79
XM_017022738.1 XP_016878227.1 26 - 27 3293 79
XM_017022739.1 XP_016878228.1 26 - 27 3317 79
XM_017022740.1 XP_016878229.1 26 - 27 3356 79
XM_017022741.1 XP_016878230.1 25 - 26 3569 79
XM_017022742.1 XP_016878231.1 26 - 27 3361 79
XM_017022743.1 XP_016878232.1 25 - 26 3227 79
XM_017022744.1 XP_016878233.1 25 - 26 3168 79
XM_017022745.1 XP_016878234.1 26 - 27 3332 79
XM_017022746.1 XP_016878235.1 25 - 26 3203 79
XM_017022748.1 XP_016878237.1 25 - 26 3569 79
XM_017022749.1 XP_016878238.1 26 - 27 3329 79
XM_017022750.1 XP_016878239.1 25 - 26 3170 79
XM_017022751.1 XP_016878240.1 25 - 26 3123 79
XM_017022752.1 XP_016878241.1 26 - 27 3229 79
XM_017022753.1 XP_016878242.1 25 - 26 3203 79
XM_017022758.1 XP_016878247.1 20 - 21 2513 79
XM_017022760.1 XP_016878249.1 25 - 26 4701 79
GenBank mRNA AF502586.1 - 25 - 26 3143 79
AF502588.1 - 25 - 26 3143 79
AF502589.1 - 24 - 25 2872 79
AF543190.1 - 24 - 25 2822 79
AK307438.1 - 26 - 27 3217 79
BC035611.1 - 5 - 6 556 79
BC057395.1 - 25 - 26 3211 79
BX538022.1 - 25 - 26 3199 79
BX647814.1 - 26 - 27 3205 79
BX647870.1 - 3 - 4 361 79

View Full Product Details