Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRG1
Entrez Gene ID
2483
Gene Name
FSHD region gene 1
Gene Aliases
FRG1A, FSG1
Chromosome Location
Chr.4: 189940819 - 189963204 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004477.2 NP_004468.1 3 - 4 444 154
GenBank mRNA AK291890.1 - 3 - 4 418 154
BC053997.1 - 3 - 4 401 154
BG426031.1 - - 475 154
L76159.1 - 3 - 4 444 154

Gene Symbol
LOC102723780
Entrez Gene ID
102723780
Gene Name
protein FRG1-like
Gene Aliases
-
Chromosome Location
Chr.22: 10940597 - 10961529 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_132320.1 4 - 5 537 154

View Full Product Details