The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027917.1 | 1 - 2 | 85 | 140 | |
GenBank mRNA | AJ851862.1 | - | 1 - 2 | 85 | 140 |