Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CDK11A
- Entrez Gene ID
-
728642
- Gene Name
- cyclin dependent kinase 11A
- Gene Aliases
-
CDC2L2, CDC2L3, CDK11-p110, CDK11-p46, CDK11-p58, PITSLRE, p58GTA
- Chromosome Location
-
Chr.1: 1702383 - 1724565 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF067524.1 |
- |
2 - 3 |
238 |
173 |
View Full Product Details