Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- WASH7P
- Entrez Gene ID
-
653635
- Gene Name
- WAS protein family homolog 7 pseudogene
- Gene Aliases
-
FAM39F, WASH5P
- Chromosome Location
-
Chr.1: 14362 - 29370 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024540.1 |
|
11 - 11 |
|
151 |
GenBank mRNA |
BU616084.1 |
- |
1 - 1 |
93 |
152 |
- Gene Symbol
- WASH3P
- Entrez Gene ID
-
374666
- Gene Name
- WAS protein family homolog 3 pseudogene
- Gene Aliases
-
FAM39DP
- Chromosome Location
-
Chr.15: 101960813 - 101976605 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003659.2 |
|
11 - 11 |
|
156 |
GenBank mRNA |
AK097269.1 |
- |
3 - 3 |
|
156 |
BQ937971.1 |
- |
1 - 1 |
|
156 |
View Full Product Details