Product Details

Assay Reference Genome
Location

Chr.15:42728705 on build GRCh38
Cytoband
15q15.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
146059
Gene Symbol
CDAN1
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
Location
Chr.15:42723557-42737219 on build GRCh38
Assay Gene Location
Within Exon 20
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CDAN1 NM_138477.2 20 2868 NP_612486.2
XM_005254176.4 20 2783 XP_005254233.1
XM_011521270.2 20 2808 XP_011519572.1
XM_011521271.2 20 2805 XP_011519573.1
XM_011521274.2 17 1854 XP_011519576.1
AF525398.1 20 2865 AAO14994.1
BC001092.2 12 1323 AAH01092.1
BC008333.1 12 1323 AAH08333.1
BC008334.1 12 1323 AAH08334.1
BC052568.1 20 2756 AAH52568.1
BC066640.1 20 2756 AAH66640.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv528725 Chr15:42463357 - 42812208 on Build GRCh38 Gain LRRC57 STARD9 TTBK2 SNAP23 ZNF106 CDAN1 HAUS2
nsv952615 Chr15:42714903 - 42736802 on Build GRCh38 Deletion STARD9 CDAN1
nsv457122 Chr15:42656664 - 42770350 on Build GRCh38 Loss STARD9 TTBK2 CDAN1
nsv517553 Chr15:42678701 - 42731284 on Build GRCh38 Gain+Loss STARD9 CDAN1

View Full Product Details