Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- COMT
- Entrez Gene ID
-
1312
- Gene Name
- catechol-O-methyltransferase
- Gene Aliases
-
HEL-S-98n
- Chromosome Location
-
Chr.22: 19941740 - 19969975 on Build GRCh38
- Gene Symbol
- ARVCF
- Entrez Gene ID
-
421
- Gene Name
- armadillo repeat gene deleted in velocardiofacial syndrome
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 19966491 - 20016802 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL390148.1 |
- |
1 - 1 |
|
135 |
View Full Product Details