The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001072.3 | NP_001063.2 | 1 - 1 | 789 | 127 |
GenBank mRNA | AY435141.1 | - | 1 - 1 | 650 | 127 |
BC019861.1 | - | 1 - 1 | 684 | 127 | |
DQ364250.1 | - | 1 - 1 | 658 | 127 |