The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077469.2 | NP_001070937.1 | 5 - 5 | 681 | 100 |
NM_001077470.2 | NP_001070938.1 | 4 - 4 | 541 | 100 | |
NM_001077471.2 | NP_001070939.1 | 5 - 5 | 681 | 100 | |
NM_001077472.2 | NP_001070940.1 | 4 - 4 | 541 | 100 | |
NM_001077473.2 | NP_001070941.1 | 4 - 4 | 541 | 100 | |
NM_001077474.2 | NP_001070942.1 | 5 - 5 | 681 | 100 | |
NM_001077475.2 | NP_001070943.1 | 4 - 4 | 541 | 100 | |
NM_001077476.2 | NP_001070944.1 | 4 - 4 | 541 | 100 | |
NM_001077477.2 | NP_001070945.1 | 4 - 4 | 541 | 100 | |
NM_001077478.2 | NP_001070946.1 | 5 - 5 | 681 | 100 | |
NM_001077479.2 | NP_001070947.1 | 4 - 4 | 541 | 100 | |
NM_001077480.2 | NP_001070948.1 | 5 - 5 | 681 | 100 | |
NM_001077481.2 | NP_001070949.1 | 5 - 5 | 681 | 100 | |
NM_001077482.2 | NP_001070950.1 | 5 - 5 | 681 | 100 | |
NM_005122.4 | NP_005113.1 | 5 - 5 | 681 | 100 | |
XM_005245693.4 | XP_005245750.1 | 4 - 4 | 954 | 100 | |
XM_005245694.4 | XP_005245751.1 | 4 - 4 | 953 | 100 | |
XM_005245697.4 | XP_005245754.1 | 5 - 5 | 564 | 100 | |
XM_011510237.2 | XP_011508539.1 | 4 - 4 | 951 | 100 | |
GenBank mRNA | AB307702.1 | - | 4 - 4 | 478 | 100 |
AK303757.1 | - | 4 - 4 | 534 | 100 | |
AK303796.1 | - | 5 - 5 | 715 | 100 | |
AK310773.1 | - | 3 - 3 | 252 | 100 | |
AK316208.1 | - | 5 - 5 | 673 | 100 | |
AY572806.1 | - | 5 - 5 | 572 | 100 | |
AY572807.1 | - | 5 - 5 | 501 | 100 | |
AY572808.1 | - | 4 - 4 | 432 | 100 | |
AY572809.1 | - | 3 - 3 | 262 | 100 | |
AY572810.1 | - | 5 - 5 | 572 | 100 | |
AY572811.1 | - | 4 - 4 | 432 | 100 | |
AY572812.1 | - | 4 - 4 | 432 | 100 | |
AY572813.1 | - | 5 - 5 | 572 | 100 | |
AY572814.1 | - | 3 - 3 | 262 | 100 | |
AY572815.1 | - | 3 - 3 | 262 | 100 | |
AY572816.1 | - | 4 - 4 | 432 | 100 | |
AY572817.1 | - | 4 - 4 | 432 | 100 | |
AY572818.1 | - | 4 - 4 | 432 | 100 | |
AY572819.1 | - | 5 - 5 | 572 | 100 | |
AY572820.1 | - | 4 - 4 | 342 | 100 | |
AY572826.1 | - | 4 - 4 | 402 | 100 | |
AY572827.1 | - | 4 - 4 | 432 | 100 | |
BC069626.1 | - | 5 - 5 | 634 | 100 | |
BC121120.1 | - | 5 - 5 | 582 | 100 | |
BC121121.1 | - | 5 - 5 | 582 | 100 | |
DQ022681.1 | - | 4 - 4 | 478 | 100 | |
HQ692838.1 | - | 4 - 4 | 478 | 100 | |
HQ692839.1 | - | 4 - 4 | 478 | 100 | |
HQ692840.1 | - | 4 - 4 | 478 | 100 | |
HQ692841.1 | - | 4 - 4 | 478 | 100 | |
HQ709178.1 | - | 4 - 4 | 478 | 100 |