Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP46A1
Entrez Gene ID
10858
Gene Name
cytochrome P450 family 46 subfamily A member 1
Gene Aliases
CP46, CYP46
Chromosome Location
Chr.14: 99684262 - 99727321 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006668.1 NP_006659.1 6 - 6 533 67
XM_005267274.4 XP_005267331.1 2 - 2 197 67
XM_011536364.1 XP_011534666.1 6 - 6 689 67
XM_011536365.1 XP_011534667.1 2 - 2 814 67
XM_017020933.1 XP_016876422.1 7 - 7 732 67
GenBank mRNA AB209749.1 - 7 - 7 690 67
AF094480.1 - 6 - 6 533 67
AK090886.1 - 1 - 1 740 67
AK295216.1 - 6 - 6 495 67
BC022539.1 - 6 - 6 634 67

View Full Product Details