Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
C2orf27B
Entrez Gene ID
408029
Gene Name
chromosome 2 open reading frame 27B
Gene Aliases
-
Chromosome Location
Chr.2: 131794961 - 131801661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214461.2 NP_999626.1 3 - 3 560 92
GenBank mRNA BC043584.1 - 3 - 3 560 92

View Full Product Details