The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002163.2 | NP_002154.1 | 9 - 9 | 2440 | 63 |
XM_017023199.1 | XP_016878688.1 | 7 - 7 | 2269 | 63 | |
GenBank mRNA | AI307662.1 | - | 1 - 1 | 63 |