Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFAP2B
Entrez Gene ID
7021
Gene Name
transcription factor AP-2 beta
Gene Aliases
AP-2B, AP2-B
Chromosome Location
Chr.6: 50817692 - 50847619 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003221.3 NP_003212.2 7 - 7 1929 115
XM_011514837.2 XP_011513139.1 8 - 8 2234 115
XM_017011233.1 XP_016866722.1 10 - 10 2610 115
XM_017011234.1 XP_016866723.1 10 - 10 2063 115
XM_017011235.1 XP_016866724.1 6 - 6 1742 115
GenBank mRNA BC037225.1 - 7 - 7 1785 115

View Full Product Details