Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPS3
Entrez Gene ID
8533
Gene Name
COP9 signalosome subunit 3
Gene Aliases
CSN3, SGN3
Chromosome Location
Chr.17: 17246624 - 17281303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199125.1 NP_001186054.1 12 - 12 1424 109
NM_001316354.1 NP_001303283.1 12 - 12 1558 109
NM_001316355.1 NP_001303284.1 11 - 11 1297 109
NM_001316356.1 NP_001303285.1 11 - 11 1341 109
NM_001316357.1 NP_001303286.1 11 - 11 1421 109
NM_001316358.1 NP_001303287.1 10 - 10 1291 109
NM_003653.3 NP_003644.2 12 - 12 1471 109
XM_005256837.4 XP_005256894.1 12 - 12 1490 109
XM_005256840.3 XP_005256897.1 12 - 12 1535 109
XM_005256842.4 XP_005256899.1 11 - 11 1403 109
XM_017025246.1 XP_016880735.1 11 - 11 1329 109
GenBank mRNA AF031647.1 - 12 - 12 1376 109
AF098109.1 - 12 - 12 1396 109
AK297706.1 - 12 - 12 1446 109
AK302304.1 - 12 - 12 1424 109
AK316400.1 - 12 - 12 1503 109
BC001891.1 - 12 - 12 1445 109
BQ018146.1 - 1 - 1 109
BU952436.1 - 1 - 1 109

View Full Product Details